Tomato leaves - trace elements, NIST-1573A
Tomato leaves - trace elements, NIST-1573A, 50 g
Catalog # PROONIST-1573A
Supplier: LGC Standards PROMOCHEM
Tomato leaves - trace elements, NIST-1573A
Catalog # PROONIST-1573A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
This standard material is intended for use in evaluating the reliability of analytical methods for the determination of major, minor, and trace elements in botanical materials, agricultural food products, and materials of similar matrix.
A unit consists of 50 g of dried tomato leaves.
Certifications: Traceable to NIST and SRM numbers.
Specifications
Specifications
- Pakn:50 g
- Beskrivelse:Tomato leaves - trace elements, NIST-1573A