Organic reference standards, ibuprofen, BP
Ibuprofen impurity B, Reference standard, 1,15 mL
Katalog # PROOBP880
Leverandør: LGC Standards PROMOCHEM
Organic reference standards, ibuprofen, BP
Katalog # PROOBP880
Leverandør: LGC Standards PROMOCHEM
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakn:1,15 mL
- Standard:Reference standard
- Beskrivelse:Ibuprofen impurity B
- Cat. No.:PROOBP880
- Beskrivelse:Ibuprofen impurity B
- CAS-nr:3585-49-7
- Formel:C₁₃H₁₈O₂
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



