Organic reference standards, dexamethasone impurity standard, BP
Dexamethasone impurity standard, Reference standard, 25 mg
Katalog # PROOBP811
Leverandør: LGC Standards PROMOCHEM
Organic reference standards, dexamethasone impurity standard, BP
Katalog # PROOBP811
Leverandør: LGC Standards PROMOCHEM
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakn:25 mg
- Standard:Reference standard
- Beskrivelse:Dexamethasone impurity standard
- Cat. No.:PROOBP811
- Beskrivelse:Dexamethasone impurity standard
- CAS-nr:50-02-2
- Formel:C₂₂H₂₉FO₅
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



