Organic reference standards, testosterone, BP
Testosterone, BP 1076, Reference Standard, 25 mg
Katalog # PROOBP1076
Leverandør: LGC Standards PROMOCHEM
Organic reference standards, testosterone, BP
Katalog # PROOBP1076
Leverandør: LGC Standards PROMOCHEM
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakn:25 mg
- Standard:Reference Standard
- Beskrivelse:Testosterone, BP 1076
- Cat. No.:PROOBP1076
- Beskrivelse:Testosterone
- CAS-nr:58-20-8
- Formel:C₁₉H₂₈O₂



