Human recombinant SDCBP
Leverandør: Abnova
Om denne vare
Human SDCBP full-length ORF recombinant protein with GST-tag at N-terminal.
- Host: Wheat germ (in vitro)
- Purifification: by Glutathione sepharose 4 fast flow
- Storage buffer: 50 mM Tris-HCI, 10 mM reduced Glutathione, pH = 8,0 in the elution buffer
- Preparation method: in vitro wheat germ expression system
- Quality control testing: 12,5% SDS-PAGE stained with coomassie blue
Used in Enzyme-linked immunoabsorbent assay, western blot (recombinant protein), antibody production and protein array.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Human SDCBP full-length ORF recombinant protein with GST-tag at N-terminal.
- Host: Wheat germ (in vitro)
- Purifification: by Glutathione sepharose 4 fast flow
- Storage buffer: 50 mM Tris-HCI, 10 mM reduced Glutathione, pH = 8,0 in the elution buffer
- Preparation method: in vitro wheat germ expression system
- Quality control testing: 12,5% SDS-PAGE stained with coomassie blue
Used in Enzyme-linked immunoabsorbent assay, western blot (recombinant protein), antibody production and protein array.
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- ABNOH00006386-P01
- ABNOH00006386-P012
- Protein/peptide name:
- SDCBP
- SDCBP
- Protein/peptide type:
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Molekylvægt:
- 58,8 kDA
- 58,8 kDA
- Opbevaring betingelser:
- –80 °C
- –80 °C
Specifikationer
Product Family Options
Produktinformation
- PaknTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.ABNOH00006386-P01Protein/peptide nameSDCBPProtein/peptide typeRecombinantSpeciesHumanMolekylvægt58,8 kDAOpbevaring betingelser–80 °C - Specifikationer:
More
Specifications
Cat. No.ABNOH00006386-P012Protein/peptide nameSDCBPProtein/peptide typeRecombinantSpeciesHumanMolekylvægt58,8 kDAOpbevaring betingelser–80 °C