Orders LinkContactUsLinkComponent
Mini coolers, True North®
Mini coolers, True North®
Leverandør:   HEATHROW
Mini coolers, True North®
Leverandør:  HEATHROW
Certifikater
Mini coolers, True North®
Mini coolers, True North®
3 tilgængelige muligheder nedenfor
Product Details & Documents
Protect temperature-sensitive samples at the bench or in the freezer.
  • Polycarbonate construction with rubber non-slip feet
  • Gridded and keyed one-position lids for easy sample identification
Ideal for keeping PCR® reagents, enzymes, and other biochemical reagents cool while working at the bench.
Filled with insulating gel, the mini coolers can also be used for controlled sample freezing and for longer temperature retention samples.
Coolers HEATHS120072 and HEATHS120075 are filled with a non-toxic self-insulating gel.
Bestillingsoplysninger: Versions available for 0,5 and 1,5/2,0 ml microcentrifuge tubes and cryogenic vials and a 96-place version for 0,2 ml tubes and assay plates.
Leveringsinformation: Supplied with 0,5 ml tube adapters as standard and temperature-retention graph.
Product Family Options
Produktinformation
  • Kapacitet
    Dimensioner
    Tilgængelighed
    Price
  • HEATHS120073
    True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 2,5 h temperature retention
    True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 2,5 h temperature retention
    12×0,5 - 2 ml, 3×4 array
    145×105×100 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr2,160.00
    kr2,160.00
    1 * 1 Items
       kr2,160.00
  • HEATHS120072
    True North® mini-cooler, 0 °C, gel filled lid, blue, 1 °C for 5 h temperature retention
    True North® mini-cooler, 0 °C, gel filled lid, blue, 1 °C for 5 h temperature retention
    32×0,5 - 2 ml, 4×8 array
    245×105×100 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr4,180.00
    kr4,180.00
    1 * 1 Items
       kr4,180.00
  • HEATHS120074
    True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 3,5 h temperature retention
    True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 3,5 h temperature retention
    32×0,5 - 2 ml, 4×8 array
    245×105×100 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr3,430.00
    kr3,430.00
    1 * 1 Items
       kr3,430.00