Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Phenylacetamid 99%

250 mg
Katalog # H60626.MD
Irriterende
Leverandør:   Thermo Fisher Scientific
Phenylacetamid 99%
Katalog # H60626.MD
CAS nummer:  103-81-1
Phenylacetamid 99%
Phenylacetamid 99%

250 mg

CAS nummer:  103-81-1
Price
829,00 kr
Log ind for tilgængelighed
1 * 250 mg
Product Details & Documents

Used as a pharmaceutical intermediate.

Store at 4°C.

Synonymer:  2-Phenylacetamide
  • CAS nummer:103-81-1
  • Formel:C₈H₉NO
  • MDL nummer:MFCD00059193
  • Molecular Weight:135.17 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-147-0
Specifikationer

Specifikationstestresultater

  • Assay (HPLC):
    98,5%
  • Solubility:
    Soluble in methanol
  • Storage temperature:
    4 °C

Specifikationer

  • Pakn:
    250 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.