Leak detector solution, Snoop®
Catalog # PERCN9301329
Supplier: PerkinElmer
Specifications
- Beskrivelse:Liquid leak detector, Snoop® (237 ml)
- Environmentally Preferable:
- Pakn:1
Specifications
About this item
For use with volumetric (bubble) flowmeters as well as for checking gas delivery system plumbing for leaks.
Suitable for gas chromatography (GC) applications.
Caution: Not recommended for use upstream of capillary GC systems - An electronic leak detector is preferred.