Drawer cabinets 75/66
Leverandør: TRESTON
Gå til:
Om denne vare
Drawer units 75 series designed for storing lightweight items, in environments such as light assembly, inspection, or testing laboratory. These units equipped with base for stand alone version and castors for mobile version.
- Castors equipped with brakes
- Maximum loading capacity: 60 kg per drawer
Colour: Grey
25 tilgængelige muligheder nedenfor
Product Details & Documents
Drawer units 75 series designed for storing lightweight items, in environments such as light assembly, inspection, or testing laboratory. These units equipped with base for stand alone version and castors for mobile version.
- Castors equipped with brakes
- Maximum loading capacity: 60 kg per drawer
Colour: Grey
Product Family Options
Produktinformation
- DøreB×D×HTilgængelighedPrice
- SOVE60949108Cabinet with 1 drawer of 100 mm height
- SOVE60949109Cabinet with 1 drawer of 100 mm height and lockCabinet with 1 drawer of 100 mm height and lock2 doors750×520×600 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949101Cabinet with drawers of 100 mm height
- SOVE60949102Cabinet with 3 drawers of 100 mm and 2 drawers 150 mm heightCabinet with 3 drawers of 100 mm and 2 drawers 150 mm height5 drawers750×520×660 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949103Cabinet with 2 drawers 150 mm height, 1 drawer of 100 mm and 1 drawer of 200 mm heightCabinet with 2 drawers 150 mm height, 1 drawer of 100 mm and 1 drawer of 200 mm height4 drawers750×520×660 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949105Cabinet with 4 drawers of 100 mm height and 1 drawer of 200 mm heightCabinet with 4 drawers of 100 mm height and 1 drawer of 200 mm height5 drawers750×520×660 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949106Cabinet with 2 drawers of 100 mm height and 2 drawers of 200 mm heightCabinet with 2 drawers of 100 mm height and 2 drawers of 200 mm height4 drawers750×520×660 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949107Cabinet with drawers of 200 mm height
- SOVE60949208Cabinet with 1 drawer of 100 mm height and baseCabinet with 1 drawer of 100 mm height and base2 doors750×520×700 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949209Cabinet with 1 drawer of 100 mm height, base and lockCabinet with 1 drawer of 100 mm height, base and lock2 doors750×520×700 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949308Cabinet with 1 drawer of 100 mm height and castorsCabinet with 1 drawer of 100 mm height and castors2 doors750×520×730 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949309Cabinet with 1 drawer of 100 mm height, castors and lockCabinet with 1 drawer of 100 mm height, castors and lock2 doors750×520×730 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949201Cabinet with base and drawers of 100 mm heightCabinet with base and drawers of 100 mm height6 drawers750×520×760 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949202Cabinet with 3 drawers of 100 mm, 2 drawers 150 mm height and baseCabinet with 3 drawers of 100 mm, 2 drawers 150 mm height and base5 drawers750×520×760 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949203Cabinet with 2 drawers 150 mm height, 1 drawer of 100 mm, 1 drawer of 200 mm height and baseCabinet with 2 drawers 150 mm height, 1 drawer of 100 mm, 1 drawer of 200 mm height and base4 drawers750×520×760 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949204Cabinet with base and 4 drawers of 150 mm heightCabinet with base and 4 drawers of 150 mm height4 drawers750×520×760 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949205Cabinet with 4 drawers of 100 mm height, 1 drawer of 200 mm height and baseCabinet with 4 drawers of 100 mm height, 1 drawer of 200 mm height and base5 drawers750×520×760 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949206Cabinet with 2 drawers of 100 mm height, 2 drawers of 200 mm height and baseCabinet with 2 drawers of 100 mm height, 2 drawers of 200 mm height and base4 drawers750×520×760 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949207Cabinet with base and drawers of 200 mm heightCabinet with base and drawers of 200 mm height3 drawers750×520×760 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949302Cabinet with 3 drawers of 100 mm, 2 drawers 150 mm height and castorsCabinet with 3 drawers of 100 mm, 2 drawers 150 mm height and castors5 drawers750×520×790 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949303Cabinet with 2 drawers 150 mm height, 1 drawer of 100 mm, 1 drawer of 200 mm height and castorsCabinet with 2 drawers 150 mm height, 1 drawer of 100 mm, 1 drawer of 200 mm height and castors4 drawers750×520×790 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949304Cabinet with castors and 4 drawers of 150 mm heightCabinet with castors and 4 drawers of 150 mm height4 drawers750×520×790 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949305Cabinet with 4 drawers of 100 mm height, 1 drawer of 200 mm height and castorsCabinet with 4 drawers of 100 mm height, 1 drawer of 200 mm height and castors5 drawers750×520×790 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949306Cabinet with 2 drawers of 100 mm height, 2 drawers of 200 mm height and castorsCabinet with 2 drawers of 100 mm height, 2 drawers of 200 mm height and castors4 drawers750×520×790 mmLog ind for tilgængelighed1 * 1 Items
- SOVE60949307Cabinet with casters and drawers of 200 mm heightCabinet with casters and drawers of 200 mm height3 drawers750×520×790 mmLog ind for tilgængelighed1 * 1 Items