Volumetric flask, class A, glass stopper with individual certificate, BLAUBRAND®
Catalog # BRND937266
Supplier: Brand
Specifications
- Kapacitet:5000 ml
- Beskrivelse:Volumetric Flask NS 34/35
- Pakn:1
Specifications
About this item
Borosilicate glass 3.3, with glass stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Individual certificate included