AlphaMop™ isolator cleaning tool
Isolatorrengøringshoved med to skafter og 6 moppeovertræk, 1 moppe/æske
2 skafter (12" og 18")
1 skumpude af polyester
6 moppeovertræk af polyester
2 skafter (12" og 18")
1 skumpude af polyester
6 moppeovertræk af polyester
Catalog # 129-0273
Supplier: Texwipe
Gå til:
Product Details & Documents
The ITW Texwipe® Mini AlphaMop™ isolator cleaning tool is designed to clean isolators, biosafety cabinets, glove boxes and laminar flow hoods. The mop has a low, flat profile and rounded corners. It is made from Celcon® plastic and is totally autoclavable. The swivel joint allows the user to reach difficult-to-access areas. The replaceable foam pad ensures that the mop cover conforms to the surfaces being cleaned. 129-0273 includes two handles, 12 and 18'' (30,4 and 45,7 cm), and is easily used with one hand.
- Strong and lightweight
- Easily used in hard-to-access areas; mop covers are quickly and easily changed
- Mop covers have excellent durability and a superior strength against abrasion
Specifikationer
- Beskrivelse:Isolatorrengøringshoved med to skafter og 6 moppeovertræk
- Pakket:1 moppe/æske
2 skafter (12" og 18")
1 skumpude af polyester
6 moppeovertræk af polyester
Accessories
Produktinformation
- Catalog #PakketBeskrivelseAvailabilityPrice
- 129-027510 foam pads/inner bag
Double bagged
10 bags/caseMini AlphaMop™ replacement polyester foam coversLog ind for tilgængelighed1 * 10 Items - 1.180,00 kr
Produktinformation
- Catalog #PakketBeskrivelseAvailabilityPrice
- 141-1058150 mop covers/case
25 covers/inner bag
6 inner bags/case
6 foam pads/caseMini-AlphaMop™ isolator cleaning tool, polyester replacement set (ISO Class 4-8)Log ind for tilgængelighed1 * 1 Each - 4.670,00 kr