Wipes, KIMTECH PREP* WETTASK*
KIMTECH* / WETTASK* DSX servietter, 318×305 mm
Catalog # 115-1997
Supplier: KIMBERLY CLARK
Jump to:
Product Details & Documents
This refillable wet wiping system is the ideal alternative to using a spray bottle or open bucket in combination with a rag, making it easy to use and versatile. Two different types, depending on application.
- Closed-bucket system keeps the pre-moistened wipes fresh in the container and avoids contamination
- Designed to be compatible with quaternary amine disinfectants
- Reduces exposure to chemical vapours, splashes and spills
SXX for working with solvents, very strong and absorbent rayon/polyester, no loose fibres. Solvent resistant won't break down when wet. Low lint level suitable for critical tasks.
DS/DSX for use with sanitisers/disinfectants. Leaves some disinfectant/sanitiser on the surface that has been wiped allowing it to work throughly.
Colour: White.
Specifications
- Beskrivelse:KIMTECH* / WETTASK* DSX servietter
- L×B:318×305 mm
- Seq:03
- Cat. No.:115-1997
Accessories
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 115-2076Dispenser bucket, 4,5 l, blueSXX/DSX wipesLog in for availability1 * 4 Items - 1.640,00 kr