Orders LinkContactUsLinkComponent
Cleanroom cuff tapes, M-Zone™ CuffStrip™
Cleanroom cuff tapes, M-Zone™ CuffStrip™
Supplier:   MICRONOVA
Cleanroom cuff tapes, M-Zone™ CuffStrip™
Supplier:  MICRONOVA
Certificates
Cleanroom cuff tapes, M-Zone™ CuffStrip™
Cleanroom cuff tapes, M-Zone™ CuffStrip™
2 Options Available Below
Product Details & Documents
A conformable polyethylene tape used to ensure an effective seal between the glove and garment in critical environments.
  • Heavy liner for easy separation
  • Available in 50,8 or 76,2 mm widths
  • Gamma irradiated with full lot traceability
Length: 0,46 m
Different colours available.
Packaging: Each double bagged pack contains two strips of tape.
Product Family Options
Product Information
  • Beskrivelse
    Farve
    Steril
    Bredde
    Availability
    Price
  • Rentrumstape, bestrålet
    hvid
    +
    76,2 mm
    Log in for availability
    Log in for availability
    1 * 200 Items
    1 * 200 Items - 14.310,00 kr
    14.310,00 kr
    1 * 200 Items
       14.310,00 kr
  • Rentrumstape, bestrålet
    hvid
    +
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 200 Par
    1 * 200 Par - 13.070,00 kr
    13.070,00 kr
    1 * 200 Par
       13.070,00 kr