Orders LinkContactUsLinkComponent
Alkaline detergent, RBS A 285 SOLID PF
Alkaline detergent, RBS A 285 SOLID PF
Alkaline detergent, RBS A 285 SOLID PF, 11.4 (0.3%)
Katalog # CHEPRBSA285SOLIDPF
Leverandør:   CHEMICAL
Alkaline detergent, RBS A 285 SOLID PF
Katalog # CHEPRBSA285SOLIDPF
Leverandør:  CHEMICAL
Alkaline detergent, RBS A 285 SOLID PF
Alkaline detergent, RBS A 285 SOLID PF
Alkaline detergent, RBS A 285 SOLID PF, 11.4 (0.3%)
Price
754,00 kr
Log ind for tilgængelighed
1 * 4,5 kg
Product Details & Documents
High performance alkaline detergent in powder form for use in washing machine - labwashers.
  • Phosphate-free
  • High detergency power
  • Low working concentration
RBS A 285 SOLID PF is recommended for cleaning glassware and laboratory equipment in automatic washing machine - labwashers having dispensing compartment for powdered detergent. It efficiently removes organic and biological residues, distilation residues etc.
Ordering information: Available in box of 2 × 4,5 kg. Please contact Avantor for details.
Caution: Not recommended for use on white metals.
Specifikationer
  • Beskrivelse:
    Alkaline detergent, RBS A 285 SOLID PF
  • pH-område:
    11.4 (0.3%)