Orders LinkContactUsLinkComponent
Alkaline detergent, RBS IND 470
Alkaline detergent, RBS IND 470
Alkaline detergent, RBS IND 470, 12.6 (2%)
Katalog # CHEPRBSIND470
Leverandør:   CHEMICAL
Alkaline detergent, RBS IND 470
Katalog # CHEPRBSIND470
Leverandør:  CHEMICAL
Alkaline detergent, RBS IND 470
Alkaline detergent, RBS IND 470
Alkaline detergent, RBS IND 470, 12.6 (2%)
Price
644,00 kr
Log ind for tilgængelighed
1 * 5 L
Product Details & Documents
Concentrated alkaline liquid detergent for radioactive decontamination of parts and surfaces made of glass and stainless steel.
  • Cleans and decontaminates
  • Halogen-free
  • high emulsifying and detergency power
RBS IND 470 is recommended for manual cleaning and soaking of parts, surfaces and containers made of stainless steel, glass and plastics. It is suitable for aqueous cleaning processes in laboratories and industry (nuclear power station, cyclotrons and others institutions that use to handle radioactive products).
Bestillingsoplysninger: Available in box of 4 × 5 litres. Please contact Avantor for details.
Advarsel: Not recommended for use on white metals.
Specifikationer
  • Beskrivelse:
    Alkaline detergent, RBS IND 470
  • pH-område:
    12.6 (2%)