Orders LinkContactUsLinkComponent
Neutral detergent, RBS T 230
Leverandør:   CHEMICAL
Neutral detergent, RBS T 230
Leverandør:  CHEMICAL
Certifikater
Neutral detergent, RBS T 230
Neutral detergent, RBS T 230
2 tilgængelige muligheder nedenfor
Product Details & Documents
For manual cleaning, soaking and ultrasonic cleaning of sensitive material, glassware and laboratory equipment.
  • Compatible with all materials
  • Concentrated solution
  • High emulsifying power
RBS T 230 is a neutral cleaning agent recommended for manual cleaning, soaking and ultrasonic cleaning of sensitive materials and laboratory equipment made of aluminum, zinc, and alloys that are sensitive to acids or bases. It efficiently removes organic and biological residues, distilation residues etc.
 : Available in box of 4 × 5 litres; 20 kg drum. Please contact Avantor for details.
Product Family Options
Produktinformation
  • pH-område
    Tilgængelighed
    Price
  • CHEPT230/E123
    Neutral detergent, RBS T 230
    Neutral detergent, RBS T 230
    7.3 (4%)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 L
    1 * 5 L - 600,00 kr
    600,00 kr
    1 * 5 L
       600,00 kr
  • CHEPT230/E148
    Neutral detergent, RBS T 230
    Neutral detergent, RBS T 230
    7.3 (4%)
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 20 kg
    1 * 20 kg - 1.820,00 kr
    1.820,00 kr
    1 * 20 kg
       1.820,00 kr