Orders LinkContactUsLinkComponent
Neutralizing agent, RBS NA 2
Neutralizing agent, RBS NA 2
Neutralizing agent, RBS NA 2, 1.8 (0.5%)
Katalog # CHEPRBSNA2
Leverandør:   CHEMICAL
Neutralizing agent, RBS NA 2
Katalog # CHEPRBSNA2
Leverandør:  CHEMICAL
Neutralizing agent, RBS NA 2
Neutralizing agent, RBS NA 2
Neutralizing agent, RBS NA 2, 1.8 (0.5%)
Price
819,00 kr
Log ind for tilgængelighed
1 * 5 L
Product Details & Documents
Improves the cleaning results in washing machines - labwashers.
  • Concentrated solution
  • Surfactant-free
  • Based on phosphoric acid
RBS NA 2 solution allows neutralisation and removal of residual alkaline traces and mineral salts from the cleaned items and the internal components of the washing machine. Can also be used in the pre-wash cycle in case of heavy residues.
Bestillingsoplysninger: Available in box of 4 x 5 litres.
Documents
Specifikationer
  • Beskrivelse:
    Neutralizing agent, RBS NA 2
  • pH-område:
    1.8 (0.5%)