Orders LinkContactUsLinkComponent
PIG® SpillBlocker® dike
PIG® SpillBlocker® dike
Dike, 60×3000×35 mm
Katalog # 121-0546
Leverandør:   NEW PIG
PIG® SpillBlocker® dike
Katalog # 121-0546
Leverandør:  NEW PIG
PIG® SpillBlocker® dike
PIG® SpillBlocker® dike
Dike, 60×3000×35 mm
Price
3.240,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Don't waste a big blocker on a small leak. This 3.5 cm high SpillBlocker is just right for leaky machinery until you can make repairs.
  • Fast protection for when a leak is starting to spread; just surround machine with dike to contain liquid
  • Exclusive New Pig formulation features a flexible, tacky bottom layer that seals tight to smooth surfaces and blocks liquids
  • Built-in, interlocking end joints create a liquid-proof seal and allow you to join as many sections as you need without extra parts
  • Confines and diverts fluids without absorbing them, making cleanup or reclamation simpler
  • Polyurethane material resists water, oils and many chemicals
High-visibility yellow colour draws attention to spill for increased safety. Reusable dike cleans easily with soap and water.
Advarsel: Due to the nature of softer urethane, the physical properties of these products may change over time with exposure to certain environmental conditions like temperature, humidity and UV radiation. Typical life expectancy is 5 years. Please inspect the stored product regularly to ensure it is in a usable state. For more details and for chemical compatibility, please call Technical Services.
Specifikationer
  • Beskrivelse:
    Dike
  • B×D×H:
    60×3000×35 mm