ContactUsLinkComponent
VWR® pHenomenal® Dissolved Oxygen Probe
  664-0042
undefined
VWR® pHenomenal® Dissolved Oxygen Probe
  664-0042
  • Tilslutning:
    kabel 3 m, 8 pin
  • Model:
    pHenomenal OXY11-3, galvanic
  • Beskrivelse:
    pHenomenal® OXY11-3, galvanik
  • Materiale:
    Epoxy
  • Seq:
    01
  • Pakn:
    1
 : Includes DO sensor including calibration vessel, cleaning solution, electrolyte, abrasive foil and manual.