Safety spectacles dispenser
Dispenser til beskyttelsesbriller, klar, 203×228×483 mm
Katalog # 111-0096
Leverandør: AK
Gå til:
Product Details & Documents
This acrylic dispenser is designed to store protective eyewear such as glasses or goggles.
- Pre-pierced holes on the top for wall mounting
- Simple and handy design for easy of use
Colour: Clear
Leveringsinformation: Mounting hardware not included.
Specifikationer
- Farve:klar
- Beskrivelse:Dispenser til beskyttelsesbriller
- B×D×H:203×228×483 mm
- Cat. No.:111-0096
- Materiale:Akryl
- Dispenser type:Kan monteres på væggen
- W×D×H:203×228×483 mm
- PPE type:Briller