Orders LinkContactUsLinkComponent
Ion selective electrode, ammonia
Ion selective electrode, ammonia
Ion selective electrode, gas sensing, for ammonia (NH3), 10-6 - 5×10⁻2 mol/l, 0...+50 °C, NH3 GSE, Plast, S7
Katalog # 662-2809
Leverandør:   Mettler - Toledo
Ion selective electrode, ammonia
Katalog # 662-2809
Leverandør:  Mettler - Toledo
Ion selective electrode, ammonia
Ion selective electrode, ammonia
Ion selective electrode, gas sensing, for ammonia (NH3), 10-6 - 5×10⁻2 mol/l, 0...+50 °C, NH3 GSE, Plast, S7
Price
10.930,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This gas-sensing electrode is intended for reliably measuring the ammonia content (NH₃).
  • Modular and versatile
  • Refillable electrolyte
  • Polymer matrix membrane
Specifikationer
  • Koncentration:
    10-6 - 5×10⁻2 mol/l
  • Tilslutning:
    S7
  • Model:
    NH3 GSE
  • Beskrivelse:
    Ion selective electrode, gas sensing, for ammonia (NH3)
  • Materiale:
    Plast
  • Temperatur:
    0...+50 °C
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Membransæt til ammoniakelektrode
    Membransæt til ammoniakelektrode
    662-2774
    Log ind for tilgængelighed
    1 * 1 Items - 4.930,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Electrolyte for gas-sensing ammonia electrode, 25 ml bottle
    Electrolyte for gas-sensing ammonia electrode, 25 ml bottle
    METT51340036
    1
    Log ind for tilgængelighed
    1 * 1 Items - 315,00 kr