ContactUsLinkComponent
PIG® trolley bag spill kit
PIG® trolley bag spill kit
Catalog # 121-0519
Supplier:  NEW PIG
undefined
PIG® trolley bag spill kit
Catalog # 121-0519
Supplier:  NEW PIG
Specifications
  • Beskrivelse:
    Spill kit, includes 30 mat pads, 6 socks, 12 pillows and 5 disposal bags
  • B×D×H:
    270×450×530 mm
  • Sugeevne:
    53,7 L
  • Pakn:
    1
Specifications
About this item
The PIG® trolley bag spill kit is intended for contingency planning and quick response to spills of acids, caustics and unknown liquid spills.
  • Durable vinyl bag is packed with high-quality PIG® absorbents
  • Wheeled trolley is easy to transport supplies directly to the spill site
  • Hook-and-loop closure keeps absorbents dry plus allows easy access
  • Highly visible bag with bold Spill Kit lettering for quick identification
  • Absorbents are organised for quick, easy response
PIG HazMat absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide.
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.