Orders LinkContactUsLinkComponent
Cleanroom clipboard TexWrite®
Cleanroom clipboard TexWrite®
TexWrite® clipboard, TX5835
Katalog # 811-0123
Leverandør:   Texwipe
Cleanroom clipboard TexWrite®
Katalog # 811-0123
Leverandør:  Texwipe
Cleanroom clipboard TexWrite®
Cleanroom clipboard TexWrite®
TexWrite® clipboard, TX5835
Price
4.670,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents
The polypropylene cleanroom clipboard is the ideal portable writing surface for use in critical environments. The rugged clipboard is constructed from rigid polypropylene and features a druable plastic clip mechanism. Rigid back allows use while standing. This clipboard is available for use with any TexWrite® documentation material.
  • Chemical resistant, corrosion resistant, durable construction
  • Suitable for use with 8,5×11" or A4 paper
  • Securely holds 1 to 50 sheets of cleanroom paper
  • Rounded corners, no sharp edges to damage gloves or garments
  • Cleanroom packaged
Documents
Specifikationer
  • Beskrivelse:
    TexWrite® clipboard, TX5835