Orders LinkContactUsLinkComponent
Buffer solutions, according to DIN 19266, Fluka™
Buffer solutions, according to DIN 19266, Fluka™
Buffer solutions, according to DIN 19266, Fluka™
Certificates
Buffer solutions, according to DIN 19266, Fluka™
7 Options Available Below
Product Details & Documents
Buffers with fungicide (except pH 10,012)
Certifications: According to DIN 19266.
Documents
Product Family Options
Product Information
  • pH-værdi
    pH nøjagtighed
    pH buffer type
    Referencetemperatur
    Pakning
    Pk Info
    Pakn
    Availability
    Price
  • 4,008
    Phthalat
    25 °C
    Plastic bottle
    1×1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 458,00 kr
    458,00 kr
    1 * 1 L
       458,00 kr
  • 6,865
    Fosfat
    25 °C
    Plastic bottle
    1×1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 458,00 kr
    458,00 kr
    1 * 1 L
       458,00 kr
  • 6,865
    Fosfat
    25 °C
    Plastic bottle
    6×1 L
    6 L
    Log in for availability
    Log in for availability
    1 * 6 L
    1 * 6 L - 2.400,00 kr
    2.400,00 kr
    1 * 6 L
       2.400,00 kr
  • 7,413
    Fosfat
    25 °C
    Plastic bottle
    1×1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 492,00 kr
    492,00 kr
    1 * 1 L
       492,00 kr
  • 9,180
    Borat
    25 °C
    Plastic bottle
    1×1 L
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 517,00 kr
    517,00 kr
    1 * 1 L
       517,00 kr
  • 9,180
    Borat
    25 °C
    Plastic bottle
    6×1 L
    6 L
    Log in for availability
    Log in for availability
    1 * 6 L
    1 * 6 L - 2.710,00 kr
    2.710,00 kr
    1 * 6 L
       2.710,00 kr
  • 10.012
    ±.020
    Bicarbonater
    25 °C
    Plastic bottle
    1×500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1.280,00 kr
    1.280,00 kr
    1 * 500 mL
       1.280,00 kr