Orders LinkContactUsLinkComponent
Dressing forceps, micro
Dressing forceps, micro
Dressing forceps, micro
Dressing forceps, micro
WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).
  • Resistant to corrosion
  • Anti-magnetic
  • Serrated
Product Family Options
  • Model
    Længde
  • Straight, fine
    105 mm
    1 * 1 Items
    1 * 1 Items - 613,00 kr
    613,00 kr
    1 * 1 Items
       613,00 kr
  • Curved, fine
    105 mm
    1 * 1 Items
    1 * 1 Items - 791,00 kr
    791,00 kr
    1 * 1 Items
       791,00 kr
  • Straight, with pin, fine
    105 mm
    1 * 1 Items
    1 * 1 Items - 678,00 kr
    678,00 kr
    1 * 1 Items
       678,00 kr
  • Curved, fine
    115 mm
    1 * 1 Items
    1 * 1 Items - 740,00 kr
    740,00 kr
    1 * 1 Items
       740,00 kr
  • Straight, pointed
    115 mm
    1 * 1 Items
    1 * 1 Items - 720,00 kr
    720,00 kr
    1 * 1 Items
       720,00 kr