Spare parts for sampling unit assembly
Leverandør: AGILENT HEWLETT PACKARD
Certifikater
Om denne vare
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
Absorberende materiale, Chemizorb® Granules
Leverandør: Merck
Citronsyre monohydrat ≥99%, powder, GPR RECTAPUR®
Leverandør: VWR Chemicals
Magnesiumsulfat, (dried basis) 98.0-102.0% USP
Leverandør: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Leverandør: VWR Chemicals
VWR®, Nitril handsker, NITRILE
Leverandør: VWR Collection
Handsker til renrum, nitril, Kimtech™ G3 NxT™
Leverandør: KIMBERLY CLARK
PEEK tubing
Leverandør: Restek
Yttrium standard solution, for ICP-MS, Pure Plus grade
Leverandør: PerkinElmer