Rain hat, 9920 RS
Leverandør: FRISTADS KANSAS
Om denne vare
This rain hat is made from PU-coated polyester jersey and features a fleece lining (polyester).
- Ear flaps with drawstring
- Bright colour for improved visibility
Weight: 170 g/m²
Certifications: EN 343 (3-1)
2 tilgængelige muligheder nedenfor
Product Details & Documents
This rain hat is made from PU-coated polyester jersey and features a fleece lining (polyester).
- Ear flaps with drawstring
- Bright colour for improved visibility
Weight: 170 g/m²
Certifications: EN 343 (3-1)
Documents
Specifications for Product Family
Specifikationer
- Catalog No:
- KWIN114043130M/L
- KWIN114043130XL/2X
- Model:
- 9920 RS
- 9920 RS
- Outfit applikationer:
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Køn:
- Unisex
- Unisex
- Stof:
- PU-coated polyester
- PU-coated polyester
- Vægt:
- 170 g/m²
- 170 g/m²
Specifikationer
Product Family Options
Produktinformation
- FarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.KWIN114043130M/LModel9920 RSOutfit applikationerOutdoorRisk areaRain (waterproof)KønUnisexStofPU-coated polyesterVægt170 g/m² - Specifikationer:
More
Specifications
Cat. No.KWIN114043130XL/2XModel9920 RSOutfit applikationerOutdoorRisk areaRain (waterproof)KønUnisexStofPU-coated polyesterVægt170 g/m²