Earmuffs, Classic, PW40
Leverandør: Portwest
Gå til:
Om denne vare
Classic ear protectors made from robust ABS.
- Lightweight and robust
- Excellent attenuation rate
SNR: 28 dB
Certifications: EN 352-1
2 tilgængelige muligheder nedenfor
Product Details & Documents
Classic ear protectors made from robust ABS.
- Lightweight and robust
- Excellent attenuation rate
SNR: 28 dB
Certifications: EN 352-1
Product Family Options
Produktinformation
- FarveHyldehøjdeTilgængelighedPrice
- PRTWPW40YEREarmuffs, Classic, 28 dB, yellow
- PRTWPW40REREarmuffs, Classic, 28 dB, red