ContactUsLinkComponent
Vand med 0,1% Eddikesyre, LiChrosolv®, hypergrade for LC-MS, Supelco®
  1.59007.2500
 :  Merck
 :  
Vand med 0,1% Eddikesyre, LiChrosolv®, hypergrade for LC-MS, Supelco®
  1.59007.2500
 :  Merck
 :  
  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L
  • Seq:
    5096
 :
 :100 °C (1013 hPa)
 :M-Clarity™ quality level = MQ100
 :Stue temperatur
 :7732-18-5