Orders LinkContactUsLinkComponent
Reducing solution for ORP electrodes
Reducing solution for ORP electrodes
Reducing solution for ORP electrodes, 500 ml bottle, 1
Katalog # HACH2965349
Leverandør:   Hach
Reducing solution for ORP electrodes
Katalog # HACH2965349
Leverandør:  Hach
CAS nummer:  
Reducing solution for ORP electrodes
Reducing solution for ORP electrodes
Reducing solution for ORP electrodes, 500 ml bottle, 1
CAS nummer:  
Price
426,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Used to condition ORP electrode surfaces for use in reducing sample matrices.
Documents
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Reducing solution for ORP electrodes, 500 ml bottle