Capillary columns, Elite-Cyclosil B for autosystem
Catalog # PERCN9316545
Supplier: PerkinElmer
Specifications
- Kolonne form:Kapillær
- Film thickness:0,25 µm
- Ø int.:0,25 mm
- Length:30 m
- Environmentally Preferable:
- Pakn:1
Specifications
About this item
Elite-Cyclosil B columns are ideal for general purpose chiral separations.