Orders LinkContactUsLinkComponent
Dissolved oxygen, reagent set
Dissolved oxygen, reagent set
Dissolved oxygen reagent set, including burette,
1 - 10 mg/l O2, Winkler, 300 ml, 100, 1 SET
Katalog # HACH2351400
Leverandør:   Hach
Dissolved oxygen, reagent set
Katalog # HACH2351400
Leverandør:  Hach
CAS nummer:  
Dissolved oxygen, reagent set
Dissolved oxygen, reagent set
Dissolved oxygen reagent set, including burette,
1 - 10 mg/l O2, Winkler, 300 ml, 100, 1 SET
CAS nummer:  
Price
2.240,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
For determination of dissolved oxygen by the Winkler method (azide modification) and buret titration.
Specifikationer
Specifikationer
  • Kemisk metode:
    Winkler
  • Pakn:
    1 SET
  • Beskrivelse:
    Dissolved oxygen reagent set, including burette
  • Prøvestørrelse:
    300 ml
  • Test pr. sæt:
    100
  • Område:
    <br />1 - 10 mg/l O<sub>2</sub>
Specifikationer