Orders LinkContactUsLinkComponent
Depth filter capsules, Supracap™ 100
Depth filter capsules, Supracap™ 100
Depth filter capsules, Supracap™ 100
Certifikater
Depth filter capsules, Supracap™ 100
5 tilgængelige muligheder nedenfor
Product Details & Documents
Supracap™ 100 depth filter capsules with PP housing are scalable capsules with Seitz® high performance depth filter media. They are suitable for process development and pilot scale applications.
  • Seamless linear scalability
  • Completely disposable
  • Encapsulated design reduces operator exposure to potential biohazards
Low hold-up volume allows for increased product recovery and requires low post-use rinse volumes.
Applications include biopharmaceuticals, mammalian cell cultures, therapeutic protein, vaccines, blood plasma proteins and serum, yeast, E.coli lysates and refolds.
The Seitz® Bio 20 media grade is made of highly purified natural and modified celluloses and is free from inorganic materials. Seitz® Bio 20 media is ideal for ion sensitive products such as therapeutic proteins, dialysis solutions, and protein-based diagnostics.
Seitz® P-series media are comprised of cellulose fibers, filter aids (diatomaceous earth and perlite), and resins. This grade is recommended for coarse filtration in the biotechnological and pharmaceutical industries.
Seitz® HP-series media are feature two distinct layers of Seitz® P-series depth filter sheets that feature a more permeable layer followed by a less permeable layer (PDD1, PDE2, PDH4 and PDK5). Seitz® HP-series media performs exceptionally well in low viability, high solids-containing applications, and with wider distribution of particle sizes in biotech applications.
Certifications: Manufactured under a quality management system certified to ISO 9000 and ISO 9001, and an environmental management system certified to ISO 14001.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • PALLNP5LPDE21
  • PALLNP5LPDH41
  • PALLNP5LPDH46
  • PALLNP5LP2001
  • PALLNP6PDP81
  • Indløb/udløb:
  • 1 - 1-1/2" sanitær flange
  • 13 mm enkelt slangemodhage
  • 1 - 1-1/2" sanitær flange
  • Ø×L:
  • 154×213 mm
  • 154×263 mm
  • 154×213 mm
  • Max. temperature:
  • 40 °C
  • 40 °C
  • 40 °C
  • 40 °C
  • 40 °C
  • Maks. tryk:
  • 3 bar
  • 3 bar
  • 3 bar
  • 3 bar
  • 3 bar
Specifikationer
Product Family Options
Produktinformation
  • Model
    Porestørrelse
    Filtreringsområde
    Tilgængelighed
    Price
  • Seitz® PDE2 media with 1 - 1-1/2" sanitary flange connections, 154×213 mm
    0,2 - 3,5 µm
    250 cm²
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.020,00 kr
    2.020,00 kr
    1 * 1 Items
       2.020,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    PALLNP5LPDE21
    Indløb/udløb
    Ø×L
    Max. temperature
    40 °C
    Maks. tryk
    3 bar
  • Seitz® PDH4 media with 1 - 1-1/2" sanitary flange connections, 154×213 mm
    0,5 - 15,0 µm
    250 cm²
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.650,00 kr
    2.650,00 kr
    1 * 1 Items
       2.650,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    PALLNP5LPDH41
    Indløb/udløb
    1 - 1-1/2" sanitær flange
    Ø×L
    154×213 mm
    Max. temperature
    40 °C
    Maks. tryk
    3 bar
  • Seitz® PDH4 media with 13 mm single hose barb connections, 154×263 mm
    0,5 - 15,0 µm
    250 cm²
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.650,00 kr
    2.650,00 kr
    1 * 1 Items
       2.650,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    PALLNP5LPDH46
    Indløb/udløb
    13 mm enkelt slangemodhage
    Ø×L
    154×263 mm
    Max. temperature
    40 °C
    Maks. tryk
    3 bar
  • Seitz® P200 media with 1 - 1-1/2" sanitary flange connections, 154×213 mm
    3,0 - 6,0 µm
    500 cm²
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.650,00 kr
    2.650,00 kr
    1 * 1 Items
       2.650,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    PALLNP5LP2001
    Indløb/udløb
    1 - 1-1/2" sanitær flange
    Ø×L
    154×213 mm
    Max. temperature
    40 °C
    Maks. tryk
    3 bar
  • Seitz® PDP8 media with 1 - 1-1/2" hose barb inlet/outlet
    6,0 - 30,0 µm
    1000 cm²
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 3.500,00 kr
    3.500,00 kr
    1 * 1 Items
       3.500,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    PALLNP6PDP81
    Indløb/udløb
    Ø×L
    Max. temperature
    40 °C
    Maks. tryk
    3 bar