Dog plasma
Dog (canine) plasma with sodium heparin, 100 µl
S2930-100
: Biowest
Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.
- Pakn:100 µl
- Beskrivelse:Dog (canine) plasma with sodium heparin