Orders LinkContactUsLinkComponent
Ternary tee mixer
Ternary tee mixer
Ternary tee mixer, stainless steel, 25 µl
Katalog # REST25138
Leverandør:   Restek
Ternary tee mixer
Katalog # REST25138
Leverandør:  Restek
Ternary tee mixer
Ternary tee mixer
Ternary tee mixer, stainless steel, 25 µl
Price
8.290,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This stainless steel ternary tee mixer is an efficient cross-flow shearing mechanism.
  • Reduced baseline noise
  • Increased sensitivity
  • Increased reaction efficiency in post-column derivatisation
  • Improved gradient accuracy for more reproducible results
Specifikationer
  • Beskrivelse:
    Ternary tee mixer, stainless steel, 25 µl