Orders LinkContactUsLinkComponent
Filling solution for ISE electrodes, sodium
Filling solution for ISE electrodes, sodium
Electrode filling solution, 0,02 M NH4Cl, for use with ISENA381 IntelliCAL™ electrode, 59 ml bottle, with dispenser cap, 59 mL
Katalog # HACH2965126
Leverandør:   Hach
Filling solution for ISE electrodes, sodium
Katalog # HACH2965126
Leverandør:  Hach
CAS nummer:  
Filling solution for ISE electrodes, sodium
Filling solution for ISE electrodes, sodium
Electrode filling solution, 0,02 M NH4Cl, for use with ISENA381 IntelliCAL™ electrode, 59 ml bottle, with dispenser cap, 59 mL
CAS nummer:  
Price
260,00 kr
Log ind for tilgængelighed
1 * 59 mL
Product Details & Documents
Internal filling solution for use with ISENA381 IntelliCAL™ Ultra sodium ion selective electrodes.
Specifikationer
Specifikationer
  • Pakn:
    59 mL
  • Beskrivelse:
    Electrode filling solution, 0,02 M NH4Cl, for use with ISENA381 IntelliCAL™ electrode, 59 ml bottle, with dispenser cap