Humidification chamber
Humidification chamber, Akryl
Katalog # REST24282
Leverandør: Restek
Gå til:
Product Details & Documents
It is important to use humidified air or nitrogen to help remove volatile organic contaminants when cleaning SilcoCan® or TO-Can® canisters.
- Chamber withstands pressure up to 90 psi
- Easy-to-open lid for filling with water
- Inlet and outlet compression fittings for easy connection to pressure lines on your cleaning system
It is important to use humidified air or nitrogen to help remove volatile organic contaminants when cleaning SilcoCan® or TO-Can® canisters.
Specifikationer
- Beskrivelse:Humidification chamber
- Materiale:Akryl