Orders LinkContactUsLinkComponent
Cleaned foam cartridges
Leverandør:   Restek
Cleaned foam cartridges
Leverandør:  Restek
Certifikater
Cleaned foam cartridges
Cleaned foam cartridges
2 tilgængelige muligheder nedenfor
Product Details & Documents
These precleaned cartridges are suitable for the air sampling during ambient, indoor and industrial hygiene applications.
  • Ready to use for collection
  • Made from cleaned PU foam
  • PUF/XAD-2 sandwiches for semi-volatiles
Used in monitoring semi-volatiles organic compounds (SVOC) such as pesticides, PCB, and PAH.
Product Family Options
Produktinformation
  • Materiale
    Tilgængelighed
    Price
  • REST22117
    Small XAD cartridge, 30 mm, O.D.×L: 22×100 mm
    Small XAD cartridge, 30 mm, O.D.×L: 22×100 mm
    PU-skum
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.570,00 kr
    1.570,00 kr
    1 * 1 Items
       1.570,00 kr
  • REST22116
    Small cartridge, 76 mm, O.D.×L: 22×100 mm
    Small cartridge, 76 mm, O.D.×L: 22×100 mm
    PU-skum
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 911,00 kr
    911,00 kr
    1 * 1 Items
       911,00 kr