ImmPACT™ DAB substrat
ImmPACT™ DAB substrat, 120 mL
ImmPACT™ DAB peroxidase substrate is a great choice for automated staining. It is an entirely new formula for a diaminobenzidine-based peroxidase substrate.
- ImmPACT™ DAB-brugsopløsning er stabil i mindst 5 dage ved stuetemperatur, og mindst to uger ved opbevaring i køleskab
- Anbefales til både manuelle og automatiserede farvningsmetoder
- Meget mere intens farve med alle peroxidase-detektionssystemer end andre DAB-præparater på markedet
The ImmPACT™ DAB working solution is stable for at least five days at room temperature and for at least two weeks when stored refrigerated. Recommended for both manual and automated staining methods. Much more intense colour with any peroxidase detection system than other commercially available DAB preparations.
- Pakn:120 mL
- Beskrivelse:ImmPACT™ DAB substrat