Bilirubin conjugate, ditaurate, disodium salt, Calbiochem®
Katalog # 201102-100
Leverandør: Merck Millipore (Calbiochem)
CAS nummer:
Specifikationer
- Pakn:100 mg
- Beskrivelse:Bilirubin conjugate, ditaurate, disodium salt
Specifikationer
Om denne vare
Bilirubin conjugate, ditaurate is suitable as a direct bilirubin standard. It behaves similarly to purified bilirubin glucuronides from human bile in diazo tests.
Note: Unstable in solution.