Orders LinkContactUsLinkComponent
Transfektionsreagenser, DharmaFECT® Duo
Transfektionsreagenser, DharmaFECT® Duo
Supplier:  Horizon Discovery
Certificates
4 Options Available Below
Product Details & Documents
Et lipid-baseret reagens, som er specielt sammensat til co-transfektion af plasmid og siRNA.
  • Effektiv transfektion af to separate molekyler med en enkelt formulering
  • Høj transfektionseffektivitet for både siRNA og plasmid uden at kompromittere cellens levedygtighed
  • Potent target-knockdown og plasmidekspression
  • Uden mikrobielle kontaminanter og ribonukleaser
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • DHART-2010-01
  • DHART-2010-02
  • DHART-2010-03
  • DHART-2010-04
  • DNA og siRNA:
  • Yes
  • Yes
  • Yes
  • Yes
Specifications
Product Family Options
Product Information
  • Beskrivelse
    Pakn
    Availability
    Price
  • DharmaFECT Duo, 0.2 ml
    0,2 mL
    Log in for availability
    Log in for availability
    1 * 0,2 mL
    1 * 0,2 mL
    Specifications:
    More
    Specifications
    Cat. No.
    DHART-2010-01
    DNA og siRNA
    Yes
  • DharmaFECT Duo, 0.75 ml
    0,75 mL
    Log in for availability
    Log in for availability
    1 * 0,75 mL
    1 * 0,75 mL
    Specifications:
    More
    Specifications
    Cat. No.
    DHART-2010-02
    DNA og siRNA
    Yes
  • DharmaFECT Duo, 1.5 ml
    1,5 mL
    Log in for availability
    Log in for availability
    1 * 1,5 mL
    1 * 1,5 mL
    Specifications:
    More
    Specifications
    Cat. No.
    DHART-2010-03
    DNA og siRNA
    Yes
  • DharmaFECT® Duo, 1,5 ml x 5 rør
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
    Specifications:
    More
    Specifications
    Cat. No.
    DHART-2010-04
    DNA og siRNA
    Yes