Thermal desorber tubes, unconditioned
Leverandør: Restek
Certifikater
Om denne vare
Tube made of stainless steel are durable and unconditioned.
- Each tube maintains its unique serial number which is etched for easy identication
- Each tube has an arrow indicating flow direction to reduce errors during use
- Robust barcode on tube for recording and tracking
These tubes are offered with a variety of sorbent packing materials to collect wide range of Volatile organic compounds, for many GC applications including indoor and outdoor, ambient and industrial hygiene environments.
Frequently Bought Together