Orders LinkContactUsLinkComponent
HPTLC Vario system
HPTLC Vario system
HPTLC Vario-system
Catalog # 552-0049
Supplier:   CAMAG
HPTLC Vario system
Catalog # 552-0049
Supplier:  CAMAG
HPTLC Vario system
HPTLC Vario system
HPTLC Vario-system
Price
22.710,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Time saving by optimisation of development conditions.
  • Development with six different solvents can be tested side by side
  • Both sandwich and tank configurations can be simulated side by side, making results directly comparable
  • Six different conditions of pre-equilibration, including relative humidity, can be tested simultaneously
  • These variations of developing conditions can be freely combined
Delivery information: The HPTLC Vario system comprises an HPTLC Vario chamber for 100×100 mm plates and an HPTLC scoring unit for preparation of TLC/HPTLC plates.
Specifications
  • Beskrivelse:
    HPTLC Vario-system
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 551-8558
    Scoring blades for 551-8556
    Log in for availability
    1 * 10 Items - 471,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 551-8556
    HPTLC-scoringsenhed til klargøring af TLC/HPTLC-plader
    Log in for availability
    1 * 1 Items - 9.670,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 551-8555
    HPTLC Vario-kammer til 100×100 mm plader
    Log in for availability
    1 * 1 Items - 13.240,00 kr