Inlet adaptor kit for dual column installation
REST27185
: Restek
- Beskrivelse:Inlet adaptor kit for dual column installation
- Pakn:1
Inlet adaptor kit for dual column installation ideal for Agilent capillary injectors, split/splitless fitting for capillary columns.
- Split/splitless fitting that accepts standard, two-hole capillary ferrules
- Easier to install capillary columns due to the nut protruding farther from the insulated injection port chamber
: Kit includes adaptor fitting, 1/16" capillary nut, gold-plated 1,2 mm ID dual Vespel® ring inlet seal, one 0,4 mm ID two-hole ferrule and one 0,5 mm ID two-hole ferrule.