Flexible inspection light
Flexible inspection light
Katalog # REST22627
Leverandør: Restek
Gå til:
Product Details & Documents
This flexible inspeciton light inspects inside surfaces of sample cylinders or other chambers.
- Has a reach of 355 mm
- LED life of 100000 hours
Specifikationer
- Beskrivelse:Flexible inspection light