Orders LinkContactUsLinkComponent
Fittings, secure-fit
 :   Restek
Fittings, secure-fit
 :  Restek
Fittings, secure-fit
Fittings, secure-fit
Secure-Fit provides a good connection between HPLC components for trouble-free chromatography.
  • Consistent, leak-free seal
  • Eliminates the excess dead volume
Internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while the nut is finger tightened.
Product Family Options
Single connectors
  • Beskrivelse
    Materiale
    Længde
  • Secure-fit fittings, 0,127 mm I-Ø
    Rustfrit stål
    60 mm
    1 * 1 Items
    1 * 1 Items
  • Secure-fit fittings, 0,127 mm I-Ø
    Rustfrit stål
    100 mm
    1 * 1 Items
    1 * 1 Items
  • Secure-fit fittings, 0,178 mm I-Ø
    Rustfrit stål
    300 mm
    1 * 1 Items
    1 * 1 Items - 687,00 kr
    687,00 kr
    1 * 1 Items
       687,00 kr
  • Secure-fit fittings, 0,254 mm I-Ø
    Rustfrit stål
    200 mm
    1 * 1 Items
    1 * 1 Items
  • Secure-fit fittings, 0,178 mm I-Ø
    PEEK
    200 mm
    1 * 1 Items
    1 * 1 Items - 657,00 kr
    657,00 kr
    1 * 1 Items
       657,00 kr
  • Secure-fit fittings, 0,178 mm I-Ø
    PEEK
    300 mm
    1 * 1 Items
    1 * 1 Items - 657,00 kr
    657,00 kr
    1 * 1 Items
       657,00 kr
Ferrules
  • Beskrivelse
    Materiale
    Længde
  • Ferrule for Secure-Fit, 1,6 mm
    Rustfrit stål
    1 * 10 Items
    1 * 10 Items - 1.500,00 kr
    1.500,00 kr
    1 * 10 Items
       1.500,00 kr
Two connectors
  • Beskrivelse
    Materiale
    Længde
  • Secure-fit fittings, 0,178 mm I-Ø
    Rustfrit stål
    100 mm
    1 * 1 Items
    1 * 1 Items - 1.120,00 kr
    1.120,00 kr
    1 * 1 Items
       1.120,00 kr
  • Secure-fit fittings, 0,127 mm I-Ø
    Rustfrit stål
    500 mm
    1 * 1 Items
    1 * 1 Items - 1.300,00 kr
    1.300,00 kr
    1 * 1 Items
       1.300,00 kr