Orders LinkContactUsLinkComponent
Miniature air sampling canisters
Leverandør:   Restek
Miniature air sampling canisters
Leverandør:  Restek
Certifikater
Miniature air sampling canisters
Miniature air sampling canisters
12 tilgængelige muligheder nedenfor
Product Details & Documents
These small canisters are designed for controlled sampling, as an alternative to tube and pump samplers.
  • Treated/electropolished stainless steel
  • Equipped with quick-connect fitting for sampling and analysis instruments
  • Canister with about 1000 cc suitable for sampling volatile organic compounds in air
Suitable for indoor air, personal, emergency response, or soil gas sampling.
 : NOTE: Suitable for low pressure application. Should not exceed 40 psig (2,75 bar).
Product Family Options
Electropolished stainless steel canisters
Produktinformation
  • Tilgængelighed
    Price
  • REST24188
    Miniature canister, 400 ml
    Miniature canister, 400 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr3,400.00
    kr3,400.00
    1 * 1 Items
       kr3,400.00
  • REST24191
    Miniature canister with metal-seated diaphragm valve, 400 ml
    Miniature canister with metal-seated diaphragm valve, 400 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • REST24194
    Miniature canister, 1000 ml
    Miniature canister, 1000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • REST24197
    Miniature canister with metal-seated diaphragm valve, 1000 ml
    Miniature canister with metal-seated diaphragm valve, 1000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Siltek® treated canisters
Produktinformation
  • Tilgængelighed
    Price
  • REST24189
    Miniature canister, 400 ml
    Miniature canister, 400 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr4,200.00
    kr4,200.00
    1 * 1 Items
       kr4,200.00
  • REST24190
    Miniature canister with siltek® treated fitting, 400 ml
    Miniature canister with siltek® treated fitting, 400 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr6,890.00
    kr6,890.00
    1 * 1 Items
       kr6,890.00
  • REST24192
    Miniature canister with metal-seated diaphragm valve, 400 ml
    Miniature canister with metal-seated diaphragm valve, 400 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • REST24193
    Miniature canister with siltek® treated diaphragm valve, 400 ml
    Miniature canister with siltek® treated diaphragm valve, 400 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • REST24195
    Miniature canister, 1000 ml
    Miniature canister, 1000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • REST24196
    Miniature canister with siltek® treated fitting, 1000 ml
    Miniature canister with siltek® treated fitting, 1000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • REST24198
    Miniature canister with metal-seated diaphragm valve, 1000 ml
    Miniature canister with metal-seated diaphragm valve, 1000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • REST24199
    Miniature canister with siltek® treated diaphragm valve, 1000 ml
    Miniature canister with siltek® treated diaphragm valve, 1000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Replacement parts
Produktinformation
  • Katalog #
    Beskrivelse
    Tilgængelighed
    Price
  • Quick-connect stem fitting, siltek® treated, with 6,35 mm tube fitting connection
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr0.00
    kr0.00
    1 * 1 Items
       kr0.00
Produktinformation
  • Katalog #
    Beskrivelse
    Tilgængelighed
    Price
  • Quick-connect body fitting with 6,35 mm tube fitting connection
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr0.00
    kr0.00
    1 * 1 Items
       kr0.00
Produktinformation
  • Katalog #
    Beskrivelse
    Tilgængelighed
    Price
  • Quick-connect stem fitting with 6,35 mm tube fitting connection
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr0.00
    kr0.00
    1 * 1 Items
       kr0.00