Orders LinkContactUsLinkComponent
Ion source cleaning powder
Ion source cleaning powder
Ion source cleaning powder, 1 kg
Katalog # REST22685
Leverandør:   Restek
Ion source cleaning powder
Katalog # REST22685
Leverandør:  Restek
Ion source cleaning powder
Ion source cleaning powder
Ion source cleaning powder, 1 kg
Price
448,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
This aluminum oxide powder is used to clean surfaces that contact the sample or ion beam when you encounter poor sensitivity and inadequate abundances at high masses.
Specifikationer
  • Beskrivelse:
    Ion source cleaning powder, 1 kg