Orders LinkContactUsLinkComponent
ECD vent trap
ECD vent trap
ECD vent trap with fitting size 1/8"
Katalog # REST22017
Leverandør:   Restek
ECD vent trap
Katalog # REST22017
Leverandør:  Restek
ECD vent trap
ECD vent trap
ECD vent trap with fitting size 1/8"
Price
694,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This trap reduces the release of hazardous materials from the ECD vent into the lab.
Dimensions: 152×25 mm
Leveringsinformation: Includes connecting lines and mounting kit.
Specifikationer
  • Beskrivelse:
    ECD vent trap with fitting size 1/8"