ECD vent trap
ECD vent trap with fitting size 1/8"
Katalog # REST22017
Leverandør: Restek
Gå til:
Product Details & Documents
This trap reduces the release of hazardous materials from the ECD vent into the lab.
Dimensions: 152×25 mm
Leveringsinformation: Includes connecting lines and mounting kit.
Specifikationer
- Beskrivelse:ECD vent trap with fitting size 1/8"