Orders LinkContactUsLinkComponent
Group Lock Boxes, Portable
Group Lock Boxes, Portable
Supplier:   Brady
Group Lock Boxes, Portable
Supplier:  Brady
Certificates
Group Lock Boxes, Portable
Group Lock Boxes, Portable
2 Options Available Below
Product Details & Documents
For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps.
  • Made of mild sheet steel
  • Slot in front allows keys to be inserted when box is locked
Can also be used as a lock storage box holding up to 40 padlocks.
Product Family Options
Product Information
  • Beskrivelse
    Farve
    B×D×H
    Availability
    Price
  • Group lock box, mild sheet steel, for approx. 40 padlocks
    blå
    227×89×152 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 680,00 kr
    680,00 kr
    1 * 1 Items
       680,00 kr
  • Group lock box, mild sheet steel, for approx. 40 padlocks
    rød
    227×89×152 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 680,00 kr
    680,00 kr
    1 * 1 Items
       680,00 kr